missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 310 Genetic Analyzer Buffer Vials, 4 mL

Catalog No. 401955
Encompass
Change view
Click to view available options
Quantity:
50 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
401955 50 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 401955 Supplier Applied Biosystems™ Supplier No. 401955

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Each package contains 50 vials and cap adapters

These vials for the 310 Genetic Analyzer autosampler hold buffer and water. Cap adapters are included.

  • Use with the 310 Genetic Analyzer with Windows™ OS, and septa for 0.5mL sample tubes

Specifications

Content And Storage Each package contains 50 vials and cap adapters. May be stored at ambient temperature.
For Use With (Equipment) 310 Genetic Analyzer
For Use With (Application) Sequencing
Product Type 310 Genetic Analyzer Buffer Vial
Quantity 50 Vials
Shipping Condition Room Temperature

For research use only