missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Buffer Tray for AKTA™ start

Catalog No. 361041161
Change view
Click to view available options
For Use With (Equipment):
ÄKTA Start
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
361041161 ÄKTA Start
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361041161 Supplier Cytiva Supplier No. 29024039

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Buffer bottle tray for use in the AKTA start automated protein purification system.

Specifications

Description Buffer Bottle Tray
For Use With (Equipment) ÄKTA Start