missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Starter Kit , (500mL Buffers pH 4.0, 7.0, and 10.0 and 125mL Potassium Chloride, 4M), Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Buffer Starter Kit |
| Name Note | (500mL Buffers pH 4.0, 7.0, and 10.0 and 125mL Potassium Chloride, 4M) |
| Density | 1g/mL |
| Physical Form | Liquid |
| Quantity | 1 Kit |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Specific Gravity | 1g/mL |
| Traceability to NIST | Traceable to NIST |
| Solubility Information | Soluble in water |
| Show More |