missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 9.00 (Certified), Fisher Chemical™

Catalog No. p-213088
Change view
Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Poly Bottle
PolyPac™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB114500 500 mL Poly Bottle
SB1141 1 L Poly Bottle
SB11420 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SB114500 Supplier Thermo Fisher Scientific Supplier No. BPSB114500
Only null left

Boric Acid/Potassium Chloride/Sodium Hydroxide

Specifications

Chemical Name or Material Buffer Solution, pH 9.00
Melting Point 0°C
CAS 7447-40-7, 10043-35-3, 1310-73-2, 64-02-8, 7732-18-5
Boiling Point 100°C
pH 9.00
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
MDL Number MFCD00146206
Packaging Poly Bottle
Traceability to NIST Traceable to NIST.
Grade Certified
Show More Show Less