missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 9.0 at 25°C, Certified, LabChem™

Catalog No. LC124406
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer™
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC124406 10 L Cubitainer™
LC124401 500 mL Poly Bottle
LC124402 1 L Poly Bottle
LC124404 4 L Poly Bottle
LC124405 20 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC124406 Supplier LabChem Supplier No. LC124406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Weight: 74.55 g/mol

Specifications

Chemical Name or Material Buffer Solution pH 9.0 at 25°C
Color Colorless
CAS Max % 99.27, 0.37, 0.31, 0.05
CAS 7447-40-7, 10043-35-3, 1310-73-2
Decomposition Information Boron; Hydrogen chloride
For Use With (Application) pH Calibration
pH 9.00 ±0.01
Physical Form Liquid
Quantity 10 L
Solution Type pH Buffer
Identification Passes Test
Linear Formula KCl
Packaging Cubitainer™
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 74.55
Grade Certified
Show More Show Less