missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 8.0 at 25°C, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Buffer Solution pH 8.0 at 25°C |
| Color | Colorless |
| CAS Max % | 99.2, 0.68, 0.12 |
| CAS | 7778-77-0, 1310-73-2 |
| Decomposition Information | Phosphorus oxides; Sodium oxide |
| pH | 8.00 ±0.01 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Identification | Passes Test |
| Linear Formula | KH2PO4 |
| Show More |