missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 7, BAKER ANALYZED™, Yellow, J.T. Baker™

Catalog No. 02002486
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02002486 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02002486 Supplier Avantor Supplier No. 565603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphate buffer solution for laboratory, research, or manufacturing use.

Specifications

Chemical Name or Material Buffer, pH 7 Reference standard solution
Color Yellow
For Use With (Application) pH Calibration
pH 6.96 to 7.04
Physical Form Liquid
Quantity 4 L
Solution Type pH Buffer
Flash Point 0°C (32°F)
Packaging Cubetainer
Grade ACS