missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 7.7 at 25°C, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Buffer Solution pH 7.7 at 25°C |
| Color | Colorless |
| CAS Max % | 99.21, 0.68, 0.11 |
| CAS | 7778-77-0, 1310-73-2 |
| Decomposition Information | Phosphorus oxides; Sodium oxide |
| For Use With (Application) | pH Calibration |
| pH | 7.70 ±0.01 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Solution Type | pH Buffer |
| Show More |