missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 7.40 (Certified), Fisher Chemical™

Catalog No. p-213072
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB110500 500 mL
SB1101 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB110500 Supplier Thermo Fisher Scientific Supplier No. BPSB110500
Only null left

Potassium Phosphate Monobasic/Sodium Hydroxide

Specifications

Chemical Name or Material Buffer Solution, pH 7.40
Melting Point 0°C
CAS 7778-77-0, 64-02-8, 7732-18-5, 7758-11-4
Boiling Point 100°C
Density 1.0g/cm³
pH 7.4
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
MDL Number MFCD00134130
Packaging Poly Bottle
Traceability to NIST Traceable to NIST.
Grade Certified
Show More Show Less