missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 7.40 (Certified), Fisher Chemical™

Catalog No. SB110500
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB110500 500 mL
SB1101 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB110500 Supplier Thermo Fisher Scientific Supplier No. BPSB110500
  • ONLINE EXCLUSIVE PRICE
    $196.75
    $103.68 / Each

    Save $93.07

    47.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • ONLINE EXCLUSIVE PRICE
    $1,180.50
    $622.08 / Case of 6 Each

    Save $558.42

    47.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Phosphate Monobasic/Sodium Hydroxide

Specifications

Chemical Name or Material Buffer Solution, pH 7.40
Melting Point 0°C
CAS 7778-77-0, 64-02-8, 7732-18-5, 7758-11-4
Boiling Point 100°C
Density 1.0g/cm³
pH 7.4
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
MDL Number MFCD00134130
Packaging Poly Bottle
Traceability to NIST Traceable to NIST.
Grade Certified
Show More Show Less