missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 7.4 at 25°C, Certified, LabChem™

Catalog No. LC123904
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC123904 4 L
LC123901 500 mL
LC123902 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC123904 Supplier LabChem Supplier No. LC123904

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Weight: 136.09 g/mol

Specifications

Chemical Name or Material Buffer Solution pH 7.4 at 25°C
Color Colorless
CAS Max % 99.22, 0.68, 0.1
CAS 7778-77-0, 1310-73-2
Decomposition Information Phosphorus oxides; Sodium oxide
For Use With (Application) pH Measurement
pH 7.40 ±0.01
Physical Form Liquid
Quantity 4 L
Solution Type pH Buffer
Identification Passes Test
Linear Formula KH2PO4
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 136.09
Grade Certified
Show More Show Less