missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 7.4 at 25°C, Certified, LabChem™

Catalog No. LC123901
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC123901 500 mL
LC123902 1 L
LC123904 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC123901 Supplier LabChem Supplier No. LC123901
Only null left

Specifications

Chemical Name or Material Buffer Solution pH 7.4 at 25°C
Color Colorless
CAS Max % 99.22, 0.68, 0.1
CAS 7778-77-0, 1310-73-2
Decomposition Information Phosphorus oxides; Sodium oxide
pH 7.40 ±0.01
Physical Form Liquid
Quantity 500 mL
Identification Passes Test
Linear Formula KH2PO4
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 136.09
Grade Certified
Show More Show Less