missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution, pH 7.00 , Color-coded yellow, Certified

Catalog No. ac611060040
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC611060040 4 L
AC611065000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC611060040 Supplier Thermo Scientific Chemicals Supplier No. 611060040
Only null left

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Buffer solution pH 7.00
Absorbance Pass Test
Name Note Color-coded yellow
CAS 7778-77-0
pH 6.99 to 7.01
Quantity 4 L
Identification Pass Test
Molecular Formula H2KO4P
MDL Number MFCD00011401 MFCD00147253
Vapor Pressure 760mmHg at 20°C
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES [K+].OP(O)([O-])=O
IUPAC Name potassium dihydrogen phosphate
Molecular Weight (g/mol) 136.08
PubChem CID 516951
ChEBI CHEBI:63036
Show More Show Less

RUO – Research Use Only