missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 7.00 (Certified), Fisher Chemical™

Catalog No. p-213063
Change view
Click to view available options
Quantity:
500 mL
1 L
10 L
20 L
Packaging:
Poly Bottle
PolyPac™
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB108500 500 mL Poly Bottle
SB1081 1 L Poly Bottle
SB10810 10 L PolyPac™
SB10820 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SB108500 Supplier Thermo Fisher Scientific Supplier No. BPSB108500
Only null left

Potassium Phosphate Monobasic/Sodium Hydroxide

Specifications

Chemical Name or Material Buffer Solution, pH 7.00
Melting Point 0°C
CAS 7778-77-0, 1310-73-2, 7732-18-5
Color Colorless
Density 1.003g/cm³
pH 7.00
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
Packaging Poly Bottle
MDL Number MFCD00134130
Traceability to NIST Traceable to NIST.
Vapor Pressure 14mmHg at 20°C
Synonym Chlorine Titrimeter Reagent
Grade Certified
Show More Show Less