missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 7.0 at 25°C, Color Coded Yellow, Certified, LabChem™

Catalog No. lc123805
Change view
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
10 L
20 L
Packaging:
Cubitainer™
Pack
Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC123805 20 L Cubitainer™
LC1238020 20 x 20 mL Pack
LC123801 500 mL Poly Bottle
LC123802 1 L Poly Bottle
LC123804C 4 L Cubitainer™
LC123804 4 L Poly Bottle
LC123806 10 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. LC123805 Supplier LabChem Supplier No. LC123805
Only null left

Specifications

Chemical Name or Material Buffer Solution pH 7.0 at 25°C
Name Note Color Coded Yellow
Color Yellow
CAS Max % 99.24, 0.68, 0.08
CAS 7778-77-0, 1310-73-2
Decomposition Information Phosphorus oxides; Sodium oxide
pH 7.00 ±0.01
Physical Form Liquid
Quantity 20 L
Identification Passes Test
Linear Formula KH2PO4
Packaging Cubitainer™
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 136.09
Grade Certified
Show More Show Less