missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 7.0 at 25°C, Certified, LabChem™

Catalog No. LC123706
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
Packaging:
Cubitainer™
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC123706 10 L Cubitainer™
LC123702 1 L Poly Bottle
LC123704 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC123706 Supplier LabChem Supplier No. LC123706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Weight: 136.09 g/mol

Specifications

Chemical Name or Material Buffer Solution pH 7.0 at 25°C
Color Colorless
CAS Max % 99.24, 0.68, 0.08
CAS 7778-77-0, 1310-73-2
Decomposition Information Phosphorus oxides; Sodium oxide
For Use With (Application) pH Calibration
pH 7.00 ±0.01
Physical Form Liquid
Quantity 10 L
Solution Type pH Buffer
Identification Passes Test
Linear Formula KH2PO4
Packaging Cubitainer™
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 136.09
Grade Certified
Show More Show Less