missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 7.0 (20°C), Potassium Dihydrogen Phosphate / Disodium Hydrogen Phosphate, Traceable to SRM from NIST, with Fungicide, Honeywell Fluka™
Potassium dihydrogen phosphate / disodium hydrogen phosphate, traceable to SRM from NIST, with fungicide.
Supplier: Honeywell-Fluka 336466X500ML
Description
Leading the Industry For Over 65 Years. Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured.
Composition per liter:
- 3.52 g Potassium dihydrogen phosphate (KH2PO4)
- 7.26 g di-Sodium hydrogen phosphate-2-hydrate (Na2HPO4*2H20)
Specifications
| Buffer solution pH 7.0 (20°C) | |
| 100°C | |
| ≤100.0000%,0.7300%,0.3500% | |
| 1 g/cm3 | |
| Buffer | |
| Poly Bottle |
| Potassium dihydrogen phosphate / disodium hydrogen phosphate, traceable to SRM from NIST, with fungicide | |
| ≥90.0000% | |
| 7558-79-4,7778-77-0 | |
| 6 x 500 mL | |
| MFCD00146206 | |
| Potassium sodium phosphate buffer solution |
Chemical Identifiers
| Potassium sodium phosphate buffer solution |
Safety and Handling
ShelfLife : 1260 days from date of manufacture
EINECSNumber : 231-791-2
Recommended Storage : Room Temp