missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 6.00 (Certified), Fisher Chemical™

Catalog No. SB1041
Change view
Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Poly Bottle
PolyPac™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB1041 1 L Poly Bottle
SB104500 500 mL Poly Bottle
SB10420 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SB1041 Supplier Thermo Fisher Scientific Supplier No. FLSB1041
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer Solution, pH 6.00
Melting Point 0°C
CAS 7778-77-0, 7732-18-5, 1310-73-2
Boiling Point 100°C
Color Undesignated
pH 6.00
Physical Form Liquid
Quantity 1 L
Identification Pass Test
MDL Number MFCD00134130
Packaging Poly Bottle
Traceability to NIST Traceable to NIST.
Grade Certified
Show More Show Less