missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 5.00 (Certified), Fisher Chemical™

Catalog No. SB1021
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB1021 1 L
SB102500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB1021 Supplier Thermo Fisher Scientific Supplier No. BPSB1021
Only null left
Edge

Specifications

Chemical Name or Material Buffer Solution, pH 5.00
Melting Point 0°C
CAS 877-24-7, 1310-73-2, 7732-18-5
Boiling Point 100°C
pH 5.0
Physical Form Liquid
Quantity 1 L
Identification Pass Test
MDL Number MFCD00146206
Packaging Poly Bottle
Traceability to NIST Traceable to NIST.
Grade Certified
Show More Show Less