missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 4.01 ± 0.01 at 25°C, Reference Standard, Spectrum™ Chemical
Specifications
Specifications
| Chemical Name or Material | Buffer Solution, pH 4.01 +/- 0.01 @ 25 DEG C, Reference Standard |
| CAS | 7647-01-0, 7732-18-5, 877-24-7 |
| For Use With (Application) | pH Measurement |
| For Use With (Equipment) | pH Meter |
| pH | 4.01 ±0.01 |
| Physical Form | Liquid |
| Quantity | 20 L |
| Solution Type | pH Buffer |
| Packaging | Cubitainer |
| Grade | Not Available |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.