missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 4.00, Color-Coded Red (Certified), Fisher Chemical™
Specifications
Specifications
| Chemical Name or Material | Buffer Solution, pH 4.00, Color-Coded Red |
| Absorbance | Pass Test |
| Melting Point | 0°C |
| CAS | 50-00-0, 67-56-1, 877-24-7, 7732-18-5, 16423-68-0 |
| Boiling Point | 100°C |
| Color | Red |
| Density | 1.0g/cm³ |
| Description | pH/temperature conversions given on label. |
| pH | 4.0 |
| Physical Form | Liquid |
| Show More |