missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 4.0 at 25°C, Certified, LabChem™

Catalog No. LC122705
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer™
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC122705 20 L Cubitainer™
LC122701 500 mL Poly Bottle
LC122702 1 L Poly Bottle
LC122704 4 L Poly Bottle
LC122706 10 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC122705 Supplier LabChem Supplier No. LC122705

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Weight: 204.23 g/mol

Specifications

Chemical Name or Material Buffer Solution pH 4.0 at 25°C
Color Colorless
CAS Max % 98.935, 1.02, 0.04, 0.005
CAS 877-24-7, 50-00-0, 67-56-1
Decomposition Information Formaldehyde; Carbon monoxide; Carbon dioxide
Density 1g/mL
For Use With (Application) pH Calibration
For Use With (Equipment) pH Meter
pH 4.00 to 4.01
Physical Form Liquid
Quantity 20 L
Solution Type pH Buffer
Identification Passes Test
Linear Formula KHC8H4O4
Packaging Cubitainer™
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 204.23
Grade Certified
Show More Show Less