missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 3.0 at 25°C, Certified, LabChem™

Catalog No. LC122501
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
Packaging:
Cubitainer™
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC122501 500 mL Poly Bottle
LC122502 1 L Poly Bottle
LC122504 4 L Poly Bottle
LC122506 10 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. LC122501 Supplier LabChem Supplier No. LC122501

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer Solution pH 3.0 at 25°C
Color Colorless
CAS Max % 98.785, 1.02, 0.15, 0.04, 0.005
CAS 877-24-7, 7647-01-0, 50-00-0, 67-56-1
Decomposition Information Formaldehyde; Carbon monoxide; Carbon dioxide
Density 1g/mL
For Use With (Application) pH Calibration
For Use With (Equipment) pH Meter
pH 3.00 ±0.01
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Identification Passes Test
Linear Formula KHC8H4O4
Packaging Poly Bottle
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 204.23
Grade Certified
Show More Show Less