missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 2.00 +/-0.02 at 25°C, Reference Standard, Spectrum™ Chemical

Catalog No. 18601128
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601128 1 L Poly Bottle
18601125 500 mL Poly Bottle
18601126 4 L Poly Bottle
18601127 20 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601128 Supplier Spectrum Chemical Mfg Cor Supplier No. B2251LT
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

B-225

Spectrum™ Chemical Buffer Solution, pH 2.00 ± 0.02 at 25°C, Reference Standard is a mixture of weak acid and its conjugate base in order to prevent changes in the pH of a solution. Buffer solutions let you calibrate any pH tester to ensure accurate readings when measuring pH in water samples and process liquids.

Specifications

CAS 7647-01-0, 7447-40-7, 7732-18-5
CAS Max % 1%, 98%
For Use With (Application) pH Measurement
For Use With (Equipment) pH Meter
pH 2.00 ±0.02
Physical Form Aqueous Solution
Quantity 1 L
Solution Type pH Buffer
Packaging Poly Bottle
Grade Not Available