missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 2.0 at 25°C, Certified, LabChem™

Supplier:  LabChem LC122202

 View more versions of this product

Catalog No. LC122202


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Buffer Solution pH 2.0 at 25°C
99.59,0.37,0.04
Hydrogen chloride
2.00 ±0.01
1 L
KCl
1g/mL
Soluble in water
Certified
Colorless
7447-40-7,7647-01-0
1g/mL
Liquid
Passes Test
Poly Bottle
Traceable to NIST
74.55
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions