missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 2.0 at 25°C, Certified, LabChem™
Supplier: LabChem LC122201
Specifications
| Buffer Solution pH 2.0 at 25°C | |
| 99.59,0.37,0.04 | |
| Hydrogen chloride | |
| 2.00 ±0.01 | |
| 500 mL | |
| KCl | |
| 1g/mL | |
| Soluble in water | |
| Certified |
| Colorless | |
| 7447-40-7,7647-01-0 | |
| 1g/mL | |
| Liquid | |
| Passes Test | |
| Poly Bottle | |
| Traceable to NIST | |
| 74.55 |