missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 11.0 at 25°C, Certified, LabChem™

Catalog No. LC125404
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC125404 4 L
LC125401 500 mL
LC125402 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC125404 Supplier LabChem Supplier No. LC125404

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Weight: 141.96 g/mol

Specifications

Chemical Name or Material Buffer Solution pH 11.0 at 25°C
Color Colorless
CAS Max % 99.64, 0.35, 0.01
CAS 7558-79-4, 1310-73-2
Decomposition Information Phosphine; Phosphorus oxides; Phosgene
Density 1g/mL
For Use With (Application) pH Calibration
pH 11.00 ±0.05
Physical Form Liquid
Quantity 4 L
Solution Type pH Buffer
Identification Passes Test
Linear Formula Na2HPO4
Packaging Poly Bottle
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 141.96
Grade Certified
Show More Show Less