missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 10.00, Color-Coded Blue, Mettler Toledo™

Catalog No. 01917119
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01917119 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01917119 Supplier METTLER TOLEDO Supplier No. 63056129
Only null left

METTLER TOLEDO™ solutions for electrode calibration and care.

  • Quality solutions for accurate results
  • Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions
  • Each standard solution comes with a certificate of analysis, providing traceability to NIST standards

Specifications

Chemical Name or Material Buffer Solution, pH 10.00
Melting Point 0°C
Boiling Point 100°C
Color Blue
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 10
Physical Form Liquid
Quantity 500 mL
Solution Type Calibration