missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 10.00, Color-Coded Blue (Certified), Fisher Chemical™

Catalog No. SB115500
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB115500 500 mL Poly Bottle
SB1154 4 L Poly Bottle
SB11520 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SB115500 Supplier Thermo Fisher Scientific Supplier No. BPSB115500
Edge

Specifications

Chemical Name or Material Buffer Solution, pH 10.00, Color-Coded Blue
Absorbance Pass Test
Melting Point 0°C
Boiling Point 100°C
Color Blue
CAS 6381-92-6, 584-08-7, 1332-77-0, 1310-58-3
Description Potassium Carbonate/Potassium Tetraborate/ Potassium Hydroxide/Disodium EDTA Dihydrate. pH/temperature conversions given on label.
pH 10.0
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
MDL Number MFCD00146206
Packaging Poly Bottle
Specific Gravity 1.013 at 25°C
Traceability to NIST Traceable to NIST.
Grade Certified
Show More Show Less