missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 10.00 (Certified), Fisher Chemical™
Specifications
Specifications
| Chemical Name or Material | Buffer Solution, pH 10.00 |
| CAS | 6381-92-6, 584-08-7, 1310-58-3, 7732-18-5, 12228-88-5 |
| Color | Colorless |
| pH | 10 |
| Physical Form | Liquid |
| Quantity | 20 L |
| Identification | Pass Test |
| MDL Number | MFCD00146206 |
| Packaging | PolyPac™ |
| Traceability to NIST | Traceable to NIST. |
| Show More |