missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 10.00 (Certified), Fisher Chemical™

Catalog No. SB116500
Change view
Click to view available options
Quantity:
500 mL
1 L
10 L
20 L
Packaging:
Poly Bottle
PolyPac™
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB116500 500 mL Poly Bottle
SB1161 1 L Poly Bottle
SB11610 10 L PolyPac™
SB11620 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SB116500 Supplier Thermo Fisher Scientific Supplier No. BPSB116500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Carbonate/Potassium Tetraborate/Potassium Hydroxide/Disodium EDTA Dihydrate

Specifications

Chemical Name or Material Buffer Solution, pH 10.00
CAS 6381-92-6, 584-08-7, 1310-58-3, 7732-18-5, 12228-88-5
Color Colorless
pH 10
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
MDL Number MFCD00146206
Packaging Poly Bottle
Traceability to NIST Traceable to NIST.
Grade Certified
Show More Show Less