missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 10.00 (Certified), Fisher Chemical™

Catalog No. SB1161
Change view
Click to view available options
Quantity:
500 mL
1 L
10 L
20 L
Packaging:
Poly Bottle
PolyPac™
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB1161 1 L Poly Bottle
SB116500 500 mL Poly Bottle
SB11610 10 L PolyPac™
SB11620 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SB1161 Supplier Thermo Fisher Scientific Supplier No. BPSB1161
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Potassium Carbonate/Potassium Tetraborate/Potassium Hydroxide/Disodium EDTA Dihydrate

Specifications

Chemical Name or Material Buffer Solution, pH 10.00
CAS 6381-92-6, 584-08-7, 1310-58-3, 7732-18-5, 12228-88-5
Color Colorless
pH 10
Physical Form Liquid
Quantity 1 L
Identification Pass Test
MDL Number MFCD00146206
Packaging Poly Bottle
Traceability to NIST Traceable to NIST.
Grade Certified
Show More Show Less