missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 10.00 ±0.02 at 25°C, Reference Standard, Spectrum™ Chemical

Catalog No. 18609313
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609313 20 L Poly Bottle
18609310 500 mL Poly Bottle
18609311 1 L Poly Bottle
18609312 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609313 Supplier Spectrum Chemical Mfg Cor Supplier No. B117020LTCU
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1303-96-4

Specifications

Chemical Name or Material Buffer Solution, pH 10.00 +/-0.02 @ 25 DEG C, Reference Standard
CAS 1303-96-4, 1310-73-2, 7732-18-5
For Use With (Application) pH Measurement
For Use With (Equipment) pH Meter
pH 10.00 ±0.02
Physical Form Solution
Quantity 20 L
Solution Type pH Buffer
Packaging Poly Bottle
Grade Not Available

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.