missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 1.68 +/-0.02 at 25°C, Reference Standard, Spectrum™ Chemical

B-220

Supplier:  Spectrum Chemical Mfg Cor B220500ML

 View more versions of this product

Catalog No. 18601118

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Buffer Solution, pH 1.68 ± 0.02 at 25°C, Reference Standard is an inorganic solution in water. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum's own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.

Specifications

Specifications

7647-01-0,7447-40-7,7732-18-5
Titration
1.66 to 1.70
500 mL
Poly Bottle
1%,1%,98%
pH Meter
Aqueous Solution
Standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions