missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution Concentrate, pH 4.00 ± 0.02 at 25°C, Spectrum™ Chemical

Catalog No. 18609299
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18609299 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18609299 Supplier Spectrum Chemical Mfg Cor Supplier No. B1166500MLPL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides a cost-effective, buffer solution for life sciences and pharmaceutical quality control applications when diluted as directed.

Specifications

CAS 877-24-7, 7732-18-5, 99-76-3
For Use With (Application) pH Measurement
For Use With (Equipment) pH Meter
pH 4.00 ±0.02
Physical Form Aqueous Solution
Quantity 500 mL
Solution Type Buffer
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.