missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution (Biphthalate), pH 4, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$4903.67 - $5740.44
Specifications
| Chemical Name or Material | Buffer Solution (Biphthalate), pH 4 |
|---|---|
| Specific Gravity | 1L = 1.00kg |
| Grade | BAKER ANALYZED™ Reagent |
Specifications
| Buffer Solution (Biphthalate), pH 4 | |
| 1L = 1.00kg | |
| BAKER ANALYZED™ Reagent |