missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution B, for Sulfate, Certified, LabChem™

Catalog No. LC125752
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC125752 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC125752 Supplier LabChem Supplier No. LC125752

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready-to-use buffer solution designed for sulfate determination according to APHA.

Specifications

Chemical Name or Material Buffer Solution B, for Sulfate
Color Colorless
CAS Max % 94.39, 3, 2, 0.5, 0.1
CAS 7791-18-6, 64-19-7, 6131-90-4, 7757-79-1
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
pH 3.5 ±0.5
Physical Form Liquid
Quantity 1 L
Solution Type pH Buffer
Identification Passes Test
Linear Formula MgCl2·6H2O
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 203.31
Grade Certified
Show More Show Less