missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution, 11.0, boric acid / sodium hydroxide solution / potassium chloride, Honeywell Fluka™
Boric acid / sodium hydroxide solution / potassium chloride
$170.83 - $170.83
Chemical Identifiers
| CAS | 1336-21-6 |
|---|---|
| Molecular Formula | H5NO |
| Molecular Weight (g/mol) | 35.05 |
| MDL Number | MFCD00066650 |
| InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
| PubChem CID | 14923 |
| ChEBI | CHEBI:18219 |
| IUPAC Name | azanium;hydroxide |
| SMILES | N.O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured