missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 9.18 ± 0.01 at 25°C, Ricca Chemical

Catalog No. R159500020
Change view
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Pack
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R159500020 20 x 20 mL Pack
159516 500 mL Natural Poly Bottle
361018163 1 L Natural Poly Bottle
R15950004F 4 L Cubitainer
R15951 4 L Natural Poly Bottle
R15955 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. R159500020 Supplier Ricca Chemical Company Supplier No. R159500020PK

Specifications

Chemical Name or Material Buffer, Reference Standard, pH 9.18 ± 0.01 at 25°C
Melting Point 0°C
CAS 1303-96-4, 7732-18-5
Boiling Point 100°C
CAS Min % 99.62, 0.38
Color Colorless
CAS Max % 99.62, 0.38
Density 1
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 9.18
Physical Form Liquid
Quantity 20 x 20 mL
Solution Type pH Buffer
Packaging Pack
Solubility Information Miscible in water
Show More Show Less