missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 9.18 ± 0.01 at 25°C, Ricca Chemical

Catalog No. 159516
Change view
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Pack
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
159516 500 mL Natural Poly Bottle
R159500020 20 x 20 mL Pack
361018163 1 L Natural Poly Bottle
R15950004F 4 L Cubitainer
R15951 4 L Natural Poly Bottle
R15955 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 159516 Supplier Ricca Chemical Company Supplier No. 159516

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1303-96-4

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 9.18± 0.01 at 25°C
CAS 1303-96-4, 7732-18-5
CAS Min % 98.0, 0.37
Color Colorless
CAS Max % 100.0, 0.39
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 9.18
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Packaging Natural Poly Bottle
Sterility Non-sterile
Grade Reference Standard
Show More Show Less