missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 7.38 ± 0.01 at 25°C, Ricca Chemical

Catalog No. 361018153
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361018153 1 L
15631 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361018153 Supplier Ricca Chemical Company Supplier No. 156332

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 7.38± 0.01 at 25°C
CAS 7558-79-4, 7778-77-0, 7732-18-5
CAS Min % 97.0, 0.86, 0.29
Color Colorless
CAS Max % 100.0, 0.9, 0.3
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 7.38
Physical Form Liquid
Quantity 1 L
Solution Type pH Buffer
Packaging Natural Poly Bottle
Grade ACS
Show More Show Less