missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Reference Standard, pH 7.000.01 (25°C), MilliporeSigma™ Supelco™
Specifications
Specifications
| Color | Yellow |
| Quantity | 500 mL |
| Packaging | Poly Bottle |
| MDL Number | MFCD00130682 |