missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Reference Standard, pH 4.000.01 (25°C), MilliporeSigma™ Supelco™
Specifications
Specifications
| Color | Red |
| For Use With (Application) | pH Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 4 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solution Type | pH Buffer |
| MDL Number | MFCD00130682 |
| Packaging | Poly Bottle |
| Grade | Not Available |