missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 4.00 ± 0.01 at 25°C, Ricca Chemical

Catalog No. p-2915773
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
150016 500 mL Natural Poly Bottle
150032 1 L Natural Poly Bottle
15001CT 4 L Cubitainer
15001 4 L Natural Poly Bottle
150025 10 L Cubitainer
15005 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 150016 Supplier Ricca Chemical Company Supplier No. 150016
Only null left

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 4.00 ± 0.01 at 25 deg C
CAS 877-24-7, 7732-18-5
CAS Min % 97.0, 0.99
CAS Max % 100.0, 1.03
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 4
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Packaging Natural Poly Bottle
Show More Show Less