missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 3.00 ± 0.01 at 25°C, Ricca Chemical

Catalog No. 361018129
Change view
Click to view available options
Quantity:
1 L
20 x 20 mL
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Pack
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
361018129 1 L Natural Poly Bottle
R149500020 20 x 20 mL Pack
14951 4 L Natural Poly Bottle
149525 10 L Cubitainer
14955 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 361018129 Supplier Ricca Chemical Company Supplier No. 149532

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 3.00 ± 0.01 at 25°C
CAS 877-24-7, 7732-18-5
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 3
Physical Form Liquid
Quantity 1 L
Solution Type pH Buffer
Packaging Natural Poly Bottle
Grade ACS
Show More Show Less