missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 12.45 ± 0.01 at 25°C, Ricca Chemical

Catalog No. 161832
Change view
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
Packaging:
Clear PET-G Bottle
Natural Poly Bottle
Pack
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
161832 1 L Natural Poly Bottle
R161800020 20 x 20 mL Pack
161816 500 mL Clear PET-G Bottle
13622601 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 161832 Supplier Ricca Chemical Company Supplier No. 161832

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 12.45± 0.01 at 25°C
CAS 7447-40-7, 1310-73-2, 7732-18-5
CAS Min % 97.0, 0.35, 0.27
Color Colorless
CAS Max % 100.0, 0.37, 0.28
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 12.45
Physical Form Liquid
Quantity 1 L
Solution Type pH Buffer
Packaging Natural Poly Bottle
Sterility Non-sterile
Grade Reference Standard
Show More Show Less