missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Reference Standard, pH 12.45 ± 0.01 at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company 161816
Specifications
| Buffer, Reference Standard | |
| 7447-40-7,1310-73-2,7732-18-5 | |
| Colorless Liquid | |
| 1X | |
| pH Meter and Electrode | |
| Liquid | |
| pH Buffer | |
| Non-sterile |
| pH 12.45± 0.01 at 25°C | |
| 97.0,0.35,0.27 | |
| 100.0,0.37,0.28 | |
| Calibration | |
| 12.45 | |
| 500 mL | |
| Clear PET-G Bottle | |
| Reference Standard |