missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 10.00 ± 0.01 at 25°C, Ricca Chemical

Catalog No. 361018164
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Natural Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
361018164 500 mL Natural Poly Bottle
361018165 1 L Natural Poly Bottle
16001G 4 L Amber Glass Bottle
16001CT 4 L Cubitainer
16001 4 L Natural Poly Bottle
160025 10 L Cubitainer
16005 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 361018164 Supplier Ricca Chemical Company Supplier No. 160016
Only null left

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 10.00 ± 0.01 at 25 deg C
CAS 497-19-8, 144-55-8, 1310-73-2, 7732-18-5
CAS Min % 97.0, 0.51, 0.39, 0.03
CAS Max % 100.0, 0.53, 0.41, 0.03
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 10
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Packaging Natural Poly Bottle
Show More Show Less