missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 1.68 ± 0.01 at 25°C, Fisherbrand

Catalog No. ACCU0836500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACCU0836500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ACCU0836500 Supplier Ricca Chemical Company Supplier No. ACCU0836500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer
Packaging Natural Poly Bottle
Name Note Reference Standard, pH 1.68
CAS 7447-40-7, 7732-18-5
Odor Odorless
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
Solubility Miscible with water
Solution Type Buffer Solution
Quantity 500 mL
Grade Laboratory
Show More Show Less