missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 1.68 ± 0.01 at 25°C, Ricca Chemical

Catalog No. 361018124
Change view
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Pack
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
361018124 500 mL Natural Poly Bottle
R149200020 20 x 20 mL Pack
149232 1 L Natural Poly Bottle
14921 4 L Natural Poly Bottle
149225 10 L Cubitainer
14925 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 361018124 Supplier Ricca Chemical Company Supplier No. 149216
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 1.68± 0.01 at 25°C
Melting Point 0°C
CAS 7447-40-7, 7732-18-5
Boiling Point 100°C
Color Colorless
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 1.68
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Packaging Natural Poly Bottle
Solubility Information Miscible
Grade ACS
Show More Show Less